KEY465196101 Key Organics / BIONET

3,3-dimethyl-2-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)butanoic acid

  • MDL number MFCD01315999
  • Cas Number : 338749-12-5
  • Molecular Weight : 247.294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75829

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart