KEY465196104 Key Organics / BIONET

methyl 2-[(tert-butylcarbamothioyl)amino]-3,3-dimethylbutanoate

  • MDL number MFCD01316003
  • Cas Number : 1214138-23-4
  • Molecular Weight : 260.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75843

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart