KEY465196105 Key Organics / BIONET

(2E)-3-(4-chlorophenyl)-1-(4-methoxyphenyl)prop-2-en-1-one

  • MDL number MFCD00018789
  • Cas Number : 41564-68-5
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75864

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart