KEY465196116 Key Organics / BIONET

(4E)-4-[2-(4-fluorophenyl)hydrazin-1-ylidene]-2H,3H,4H-1lambda6-thieno[2,3-b]thiopyran-1,1-dione

  • MDL number MFCD01815041
  • Cas Number : 478043-36-6
  • Molecular Weight : 310.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75885

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-313S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart