KEY465196120 Key Organics / BIONET

6-phenyl-2-(phenylsulfanyl)-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD01316222
  • Cas Number : 338749-48-7
  • Molecular Weight : 356.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75890

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-318S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart