KEY465196123 Key Organics / BIONET

(4E)-4-[2-(2,5-dichlorophenyl)hydrazin-1-ylidene]-2H,3H,4H-1lambda6-thieno[2,3-b]thiopyran-1,1-dione

  • MDL number MFCD01443936
  • Cas Number : 338749-53-4
  • Molecular Weight : 361.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75896

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-324S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart