KEY465196126 Key Organics / BIONET

(4E)-4-[2-(4-chlorophenyl)hydrazin-1-ylidene]-2H,3H,4H-1lambda6-thieno[2,3-b]thiopyran-1,1-dione

  • MDL number MFCD01443937
  • Cas Number : 338749-59-0
  • Molecular Weight : 326.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75905

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-336S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart