KEY465196129 Key Organics / BIONET

1-(furan-2-yl)pentane-1,4-dione

  • MDL number MFCD01316230
  • Cas Number : 52786-29-5
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75911

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-342S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart