KEY465196130 Key Organics / BIONET

methyl 5-cyano-2-methyl-6-{[(3-phenyl-1,2-oxazol-5-yl)methyl]amino}pyridine-3-carboxylate

  • MDL number MFCD01316233
  • Cas Number : 338749-65-8
  • Molecular Weight : 348.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75915

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-348S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart