KEY465196134 Key Organics / BIONET

5-acetyl-6-methyl-2-{[(3-phenyl-1,2-oxazol-5-yl)methyl]amino}pyridine-3-carbonitrile

  • MDL number MFCD01316235
  • Cas Number : 338749-67-0
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75919

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-352S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart