KEY465196135 Key Organics / BIONET

5-acetyl-2-({[3-(4-methoxyphenyl)-1,2-oxazol-5-yl]methyl}amino)-6-methylpyridine-3-carbonitrile

  • MDL number MFCD01316236
  • Cas Number : 338749-68-1
  • Molecular Weight : 362.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75920

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart