KEY465196138 Key Organics / BIONET

6-(4-chlorophenyl)-2-hydroxy-4-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD01316240
  • Cas Number : 147381-62-2
  • Molecular Weight : 298.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75924

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-357S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart