KEY465196142 Key Organics / BIONET

methyl 1-(3-chlorophenyl)-5-(4-chlorophenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD01315849
  • Cas Number : 318256-12-1
  • Molecular Weight : 347.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75965

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-515S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart