KEY465196159 Key Organics / BIONET

6-(3-chlorobenzoyl)-2-phenyl-5-(trifluoromethyl)-2,3-dihydropyridazin-3-one

  • MDL number MFCD02102671
  • Cas Number : 478043-61-7
  • Molecular Weight : 378.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76021

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-592S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart