KEY465196166 Key Organics / BIONET

N3-tert-butyl-N2-(4-methoxyphenyl)-1,2,3,4-tetrahydroisoquinoline-2,3-dicarboxamide

  • MDL number MFCD01316014
  • Cas Number : 318256-28-9
  • Molecular Weight : 381.476

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76084

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart