KEY465196170 Key Organics / BIONET

ethyl 2-amino-3-[(3,4,4-trifluorobut-3-en-1-yl)sulfanyl]propanoate

  • MDL number MFCD01443830
  • Cas Number : 1396965-16-4
  • Molecular Weight : 257.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76106

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-742_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart