KEY465196178 Key Organics / BIONET

methyl 2-(4-methoxybenzamido)thiophene-3-carboxylate

  • MDL number MFCD00830189
  • Cas Number : 338750-45-1
  • Molecular Weight : 291.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76134

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-816_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart