KEY465196179 Key Organics / BIONET

methyl 2-{[(1E)-2-{[(ethoxycarbonyl)amino]carbonyl}-3-oxobut-1-en-1-yl]amino}thiophene-3-carboxylate

  • MDL number MFCD01316154
  • Cas Number : 338750-48-4
  • Molecular Weight : 340.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76137

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-819_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart