KEY465196181 Key Organics / BIONET

6-amino-4-(4-methoxyphenyl)-3-methyl-1-(4-methylphenyl)-1H,4H-pyrano[2,3-c]pyrazole-5-carbonitrile

  • MDL number MFCD01239008
  • Cas Number : 300701-99-9
  • Molecular Weight : 372.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76140

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4N-822_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart