KEY465196186 Key Organics / BIONET

N-{4-[(2-hydroxy-1-phenylethyl)sulfamoyl]phenyl}acetamide

  • MDL number MFCD01316024
  • Cas Number : 338750-55-3
  • Molecular Weight : 334.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76147

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4N-904_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart