KEY465196202 Key Organics / BIONET

(3Z)-2-(methylsulfanyl)-3-{2-[3-(trifluoromethyl)phenyl]hydrazin-1-ylidene}-3H-indole

  • MDL number MFCD01815497
  • Cas Number : 478045-17-9
  • Molecular Weight : 335.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76248

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4P-375S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart