KEY465196208 Key Organics / BIONET

N-{2-[(3-cyano-4,6-dimethylpyridin-2-yl)sulfanyl]phenyl}thiophene-2-carboxamide

  • MDL number MFCD02186647
  • Cas Number : 478045-70-4
  • Molecular Weight : 365.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76314

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-647S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart