KEY465196211 Key Organics / BIONET

ethyl (4E)-2-[(2,4-dichlorophenyl)methyl]-5-(dimethylamino)-3-oxopent-4-enoate

  • MDL number MFCD05668767
  • Cas Number : 866017-66-5
  • Molecular Weight : 344.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76334

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4P-701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart