KEY465196215 Key Organics / BIONET

3-[(2,4-dichlorophenyl)methyl]-4-hydroxy-1-[(pyridin-4-yl)methyl]-1,2-dihydropyridin-2-one

  • MDL number MFCD02082740
  • Cas Number : 478045-91-9
  • Molecular Weight : 361.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4P-714_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart