KEY465196216 Key Organics / BIONET

1-[(4-chlorophenyl)methyl]-3-[(2,4-dichlorophenyl)methyl]-4-hydroxy-1,2-dihydropyridin-2-one

  • MDL number MFCD02082741
  • Cas Number : 478045-92-0
  • Molecular Weight : 394.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76345

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4P-715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart