KEY465196217 Key Organics / BIONET

N-{3-[(2,4-dichlorophenyl)methyl]-4-hydroxy-2-oxo-1,2-dihydropyridin-1-yl}-1-methyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD02082742
  • Cas Number : 478045-93-1
  • Molecular Weight : 392.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76346

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart