KEY465196224 Key Organics / BIONET

4-methoxy-N-(4H-1,2,4-triazol-4-yl)benzamide

  • MDL number MFCD00750818
  • Cas Number : 90916-77-1
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-methoxy-N-(4H-1,2,4-triazol-4-yl)benzamide
Inchi Code 1S/C10H10N4O2/c1-16-9-4-2-8(3-5-9)10(15)13-14-6-11-12-7-14/h2-7H,1H3,(H,13,15)
Inchi Key RPMGWKPGASTCEI-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76362

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0009_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart