KEY465196227 Key Organics / BIONET

4-fluoro-N-(4H-1,2,4-triazol-4-yl)benzamide

  • MDL number MFCD00448017
  • Cas Number : 331434-05-0
  • Molecular Weight : 206.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76367

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0015_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart