KEY465196229 Key Organics / BIONET

6-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl 4-methylbenzoate

  • MDL number MFCD02186462
  • Cas Number : 478046-06-9
  • Molecular Weight : 268.276

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76369

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart