KEY465196231 Key Organics / BIONET

tert-butyl 2-[(1-phenyl-1H-imidazol-2-yl)sulfanyl]acetate

  • MDL number MFCD02186516
  • Cas Number : 478046-08-1
  • Molecular Weight : 290.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76374

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0026_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart