KEY465196233 Key Organics / BIONET

methyl 4-[(1-phenyl-1H-imidazol-2-yl)sulfanyl]butanoate

  • MDL number MFCD02186517
  • Cas Number : 478046-09-2
  • Molecular Weight : 276.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76376

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0028_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart