KEY465196239 Key Organics / BIONET

6-[(3,5-dimethoxyphenyl)carbamoyl]cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD02186522
  • Cas Number : 478046-14-9
  • Molecular Weight : 305.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0041_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart