KEY465196243 Key Organics / BIONET

ethyl 2-{[(4H-1,2,4-triazol-4-yl)carbamothioyl]amino}acetate

  • MDL number MFCD02661868
  • Cas Number : 439108-24-4
  • Molecular Weight : 229.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76395

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0053_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart