KEY465196244 Key Organics / BIONET

6-benzyl-5-methyl-2-[3-(1H-pyrrol-1-yl)thiophen-2-yl]pyrazolo[1,5-a]pyrimidin-7-ol

  • MDL number MFCD02661719
  • Cas Number : 439108-25-5
  • Molecular Weight : 386.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76397

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0056_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart