KEY465196246 Key Organics / BIONET

(1R,2S)-cyclohex-4-ene-1,2-dicarboxylic acid 2-aminopyrimidin-1-ium-1-olate

  • MDL number MFCD02186826
  • Cas Number : 477712-55-3
  • Molecular Weight : 281.268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0058_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart