KEY465196250 Key Organics / BIONET

(2E)-3-[4-(3-fluoropropoxy)phenyl]-N-[2-(pyridin-3-yl)pyrimidin-4-yl]prop-2-enamide

  • MDL number MFCD02571941
  • Cas Number : 478046-38-7
  • Molecular Weight : 378.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76425

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0208_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart