KEY465196251 Key Organics / BIONET

7-{3-[(2,4-dichlorophenyl)methoxy]phenyl}pyrazolo[1,5-a]pyrimidine-3-carbonitrile

  • MDL number MFCD02571942
  • Cas Number : 478046-39-8
  • Molecular Weight : 395.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76426

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0209_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart