KEY465196256 Key Organics / BIONET

4-methyl-N-[2-(pyridin-3-yl)pyrimidin-4-yl]benzene-1-sulfonamide

  • MDL number MFCD02571943
  • Cas Number : 478046-56-9
  • Molecular Weight : 326.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76445

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0238_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart