KEY465196262 Key Organics / BIONET

7-phenyl-2-(thiophen-2-yl)-[1,2,4]triazolo[1,5-a]pyridine-8-carbonitrile

  • MDL number MFCD02661727
  • Cas Number : 439108-34-6
  • Molecular Weight : 302.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76475

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4R-0280_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart