KEY465196264 Key Organics / BIONET

4-chloro-N-[2-(pyridin-3-yl)pyrimidin-4-yl]benzene-1-sulfonamide

  • MDL number MFCD02661729
  • Cas Number : 439108-36-8
  • Molecular Weight : 346.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76477

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0283_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart