KEY465196282 Key Organics / BIONET

(3E)-1,1,1-trifluoro-4-({4-[(E)-2-phenyldiazen-1-yl]phenyl}amino)but-3-en-2-one

  • MDL number MFCD02186477
  • Cas Number : 478047-05-1
  • Molecular Weight : 319.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76523

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0602_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart