KEY465196317 Key Organics / BIONET

1-(2,4-difluorophenyl)-4-(1H-1,2,4-triazol-1-yl)-1H-pyrazol-5-ol

  • MDL number MFCD03102519
  • Cas Number : 1007014-95-0
  • Molecular Weight : 263.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76591

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-0827_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart