KEY465196331 Key Organics / BIONET

2-(benzenesulfonyl)-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)ethan-1-one

  • MDL number MFCD02187104
  • Cas Number : 300357-23-7
  • Molecular Weight : 357.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76613

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart