KEY465196347 Key Organics / BIONET

1-(4-methylbenzenesulfonyl)-N-(propan-2-yl)piperidine-4-carboxamide

  • MDL number MFCD02186811
  • Cas Number : 478047-62-0
  • Molecular Weight : 324.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76654

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1131_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart