KEY465196352 Key Organics / BIONET

methyl N-[1-(4-chlorobenzenesulfonyl)piperidin-4-yl]carbamate

  • MDL number MFCD02187401
  • Cas Number : 478047-66-4
  • Molecular Weight : 332.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76663

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1149_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart