KEY465196355 Key Organics / BIONET

N-[1-(4-chlorobenzenesulfonyl)piperidin-4-yl]pyrrolidine-1-carboxamide

  • MDL number MFCD02187404
  • Cas Number : 478047-69-7
  • Molecular Weight : 371.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76666

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1154_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart