KEY465196363 Key Organics / BIONET

11,13-dimethyl-4-sulfanyl-8-thia-3,5,10-triazatricyclo[7.4.0.0^{2,7}]trideca-1(9),2,4,6,10,12-hexaen-6-ol

  • MDL number MFCD02571123
  • Cas Number : 326891-39-8
  • Molecular Weight : 263.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76706

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1311_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart