KEY465196367 Key Organics / BIONET

[(4Z)-4,5,6,7-tetrahydro-1-benzofuran-4-ylidene]amino 4-methylbenzene-1-sulfonate

  • MDL number MFCD02187140
  • Cas Number : 180339-35-9
  • Molecular Weight : 305.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76719

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1326_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart