KEY465196378 Key Organics / BIONET

N-methyl-3-phenyl-6-(trifluoromethyl)-1,2,4-triazin-5-amine

  • MDL number MFCD03012065
  • Cas Number : 383148-46-7
  • Molecular Weight : 254.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76752

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4R-1542_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart