KEY465196389 Key Organics / BIONET

2-[(4-chlorophenyl)methylidene]-2,3-dihydro-1H-indene-1,3-dione

  • MDL number MFCD00021234
  • Cas Number : 15875-54-4
  • Molecular Weight : 268.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76834

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0281_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart